Assay Details
| Gene Symbol: jpt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089513.1 | NP_001082982.1 | 4 - 5 | 476 | 88 | |
| GenBank mRNA | BC134249.1 | - | 4 - 5 | 476 | 88 | |
More Information
Set Membership: |
|

jpt1a
Genomic Map