Assay Details
| Gene Symbol: mapkapk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101803.1 | NP_001095273.1 | 7 - 8 | 895 | 59 | |
| GenBank mRNA | AB292795.1 | - | 7 - 8 | 895 | 59 | |
More Information
Set Membership: |
|

mapkapk2b
Genomic Map