Assay Details
| Gene Symbol: mntb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103111.1 | NP_001096581.1 | 1 - 2 | 252 | 71 | |
| XM_005170271.4 | XP_005170328.1 | 1 - 2 | 277 | 71 | ||
| GenBank mRNA | BC150227.1 | - | 1 - 2 | 252 | 71 | |
More Information
Set Membership: |
|

mntb
Genomic Map