Assay Details
| Gene Symbol: nos2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104937.1 | NP_001098407.1 | 23 - 24 | 2937 | 66 | |
| XM_005165296.4 | XP_005165353.1 | 23 - 24 | 3005 | 66 | ||
| GenBank mRNA | AM084880.1 | - | 23 - 24 | 2937 | 66 | |
| AM749801.1 | - | 23 - 24 | 2937 | 66 | ||
More Information
Set Membership: |
|

nos2a
Genomic Map