Assay Details
| Gene Symbol: dlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130958.1 | NP_571033.1 | 4 - 5 | 805 | 61 | |
| GenBank mRNA | AF006488.1 | - | 4 - 5 | 805 | 61 | |
| BC076414.1 | - | 4 - 5 | 834 | 61 | ||
| CU458990.1 | - | 4 - 5 | 805 | 61 | ||
More Information
Set Membership: |
|

dlb
Genomic Map