Assay Details
| Gene Symbol: nr2f1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131180.1 | NP_571255.1 | 1 - 2 | 1147 | 83 | |
| GenBank mRNA | BC066671.1 | - | 1 - 2 | 477 | 83 | |
| X70299.1 | - | 1147 | 83 | |||
More Information
Set Membership: |
|

nr2f1a
Genomic Map