Assay Details
| Gene Symbol: nr2f1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131180.1 | NP_571255.1 | 2 - 3 | 1674 | 95 | |
| XM_005165507.4 | XP_005165564.1 | 2 - 3 | 1751 | 95 | ||
| GenBank mRNA | BC056574.1 | - | 2 - 3 | 1730 | 95 | |
| BC066671.1 | - | 2 - 3 | 1004 | 95 | ||
| X70299.1 | - | 1674 | 95 | |||
More Information
Set Membership: |
|

nr2f1a
Genomic Map