Assay Details
| Gene Symbol: pax7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF014368.1 | - | 2 - 3 | 653 | 60 | |
| AF014369.1 | - | 2 - 3 | 653 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pax7a| Gene Symbol: pax7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF014368.1 | - | 2 - 3 | 653 | 60 | |
| AF014369.1 | - | 2 - 3 | 653 | 60 | ||
Set Membership: |
|