Assay Details
| Gene Symbol: pax7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131325.2 | NP_571400.1 | 7 - 8 | 1494 | 102 | |
| NM_131332.2 | NP_571407.2 | 7 - 8 | 1494 | 102 | ||
| XM_009306286.3 | XP_009304561.1 | 7 - 8 | 1599 | 102 | ||
| GenBank mRNA | AF014368.1 | - | 7 - 8 | 1495 | 102 | |
| AF014369.1 | - | 7 - 8 | 1495 | 102 | ||
| BC163502.1 | - | 7 - 8 | 1263 | 102 | ||
| BC163523.1 | - | 7 - 8 | 1263 | 102 | ||
More Information
Set Membership: |
|

pax7a
Genomic Map