Assay Details
Gene Symbol: pax7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF014368.1 | - | 8 - 9 | 1749 | 63 | |
BC163502.1 | - | 8 - 9 | 1517 | 63 | ||
BC163523.1 | - | 8 - 9 | 1517 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
