Assay Details
| Gene Symbol: gsk3ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131390.2 | NP_571465.1 | 4 - 5 | 707 | 75 | |
| XM_009292086.3 | XP_009290361.1 | 4 - 5 | 744 | 75 | ||
| GenBank mRNA | AJ223501.1 | - | 4 - 5 | 674 | 75 | |
| BC056332.1 | - | 4 - 5 | 685 | 75 | ||
| BC065952.1 | - | 4 - 5 | 707 | 75 | ||
More Information
Set Membership: |
|

gsk3ab
Genomic Map