Assay Details
| Gene Symbol: ephb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131417.1 | NP_571492.1 | 6 - 7 | 1307 | 80 | |
| GenBank mRNA | AJ005029.1 | - | 6 - 7 | 1307 | 80 | |
| BC163275.1 | - | 6 - 7 | 1389 | 80 | ||
More Information
Set Membership: |
|

ephb4b
Genomic Map