Assay Details
| Gene Symbol: snap25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131434.1 | NP_571509.1 | 2 - 3 | 152 | 69 | |
| GenBank mRNA | AF091594.1 | - | 2 - 3 | 152 | 69 | |
| AF091595.1 | - | 1 - 2 | 197 | 69 | ||
| BC059469.1 | - | 2 - 3 | 146 | 69 | ||
More Information
Set Membership: |
|

snap25b
Genomic Map