Assay Details
| Gene Symbol: caspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131505.2 | NP_571580.1 | 1 - 2 | 292 | 101 | |
| GenBank mRNA | AF233434.1 | - | 1 - 2 | 271 | 101 | |
| BC095022.1 | - | 1 - 2 | 292 | 101 | ||
| BC164780.1 | - | 1 - 2 | 305 | 101 | ||
More Information
Set Membership: |
|

caspa
Genomic Map