Assay Details
| Gene Symbol: caspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131505.2 | NP_571580.1 | 6 - 7 | 1060 | 81 | |
| GenBank mRNA | AF233434.1 | - | 6 - 7 | 1039 | 81 | |
| BC095022.1 | - | 6 - 7 | 1060 | 81 | ||
| BC164780.1 | - | 6 - 7 | 1073 | 81 | ||
| MG957992.1 | - | 6 - 7 | 1039 | 81 | ||
More Information
Set Membership: |
|

caspa
Genomic Map