Assay Details
| Gene Symbol: pax6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131641.1 | NP_571716.1 | 6 - 7 | 840 | 77 | |
| XM_005168819.4 | XP_005168876.1 | 7 - 8 | 906 | 77 | ||
| XM_005168822.4 | XP_005168879.1 | 7 - 8 | 907 | 77 | ||
| XM_005168823.4 | XP_005168880.1 | 4 - 5 | 803 | 77 | ||
| XM_009302973.3 | XP_009301248.1 | 6 - 7 | 867 | 77 | ||
| XM_017356906.2 | XP_017212395.1 | 6 - 7 | 848 | 77 | ||
| XM_021477600.1 | XP_021333275.1 | 7 - 8 | 848 | 77 | ||
| XM_021477601.1 | XP_021333276.1 | 8 - 9 | 977 | 77 | ||
| XM_021477602.1 | XP_021333277.1 | 7 - 8 | 803 | 77 | ||
| XM_021477603.1 | XP_021333278.1 | 7 - 8 | 878 | 77 | ||
| GenBank mRNA | AF061252.1 | - | 6 - 7 | 840 | 77 | |
| BC076068.1 | - | 6 - 7 | 861 | 77 | ||
More Information
Set Membership: |
|

pax6b
Genomic Map