Assay Details
| Gene Symbol: pax6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131641.1 | NP_571716.1 | 9 - 10 | 1235 | 68 | |
| XM_005168819.4 | XP_005168876.1 | 10 - 11 | 1301 | 68 | ||
| XM_005168822.4 | XP_005168879.1 | 10 - 11 | 1302 | 68 | ||
| XM_005168823.4 | XP_005168880.1 | 7 - 8 | 1198 | 68 | ||
| XM_009302973.3 | XP_009301248.1 | 9 - 10 | 1262 | 68 | ||
| XM_017356906.2 | XP_017212395.1 | 9 - 10 | 1243 | 68 | ||
| XM_021477600.1 | XP_021333275.1 | 10 - 11 | 1243 | 68 | ||
| XM_021477601.1 | XP_021333276.1 | 11 - 12 | 1372 | 68 | ||
| XM_021477602.1 | XP_021333277.1 | 10 - 11 | 1198 | 68 | ||
| XM_021477603.1 | XP_021333278.1 | 10 - 11 | 1273 | 68 | ||
| GenBank mRNA | AF061252.1 | - | 9 - 10 | 1235 | 68 | |
More Information
Set Membership: |
|

pax6b
Genomic Map