Assay Details
| Gene Symbol: gch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131667.1 | NP_571742.1 | 3 - 4 | 610 | 81 | |
| GenBank mRNA | AJ311846.1 | - | 3 - 4 | 598 | 81 | |
| BC071298.1 | - | 3 - 4 | 610 | 81 | ||
More Information
Set Membership: |
|

gch2
Genomic Map