Assay Details
| Gene Symbol: csf1ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131672.1 | NP_571747.1 | 16 - 17 | 2436 | 72 | |
| XM_021481056.1 | XP_021336731.1 | 17 - 18 | 2566 | 72 | ||
| XM_021481057.1 | XP_021336732.1 | 17 - 18 | 2493 | 72 | ||
| GenBank mRNA | AF240639.1 | - | 16 - 17 | 2436 | 72 | |
| BC162803.1 | - | 16 - 17 | 2371 | 72 | ||
| CU458899.1 | - | 15 - 16 | 2398 | 72 | ||
More Information
Set Membership: |
|

csf1ra
Genomic Map