Assay Details
| Gene Symbol: csf1ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131672.1 | NP_571747.1 | 7 - 8 | 1346 | 91 | |
| XM_021481056.1 | XP_021336731.1 | 8 - 9 | 1476 | 91 | ||
| XM_021481057.1 | XP_021336732.1 | 8 - 9 | 1403 | 91 | ||
| GenBank mRNA | AF240639.1 | - | 7 - 8 | 1346 | 91 | |
| BC162803.1 | - | 7 - 8 | 1281 | 91 | ||
| CU458899.1 | - | 7 - 8 | 1240 | 91 | ||
More Information
Set Membership: |
|

csf1ra
Genomic Map