Assay Details
| Gene Symbol: angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131813.1 | NP_571888.1 | 8 - 9 | 1430 | 50 | |
| GenBank mRNA | AF379602.1 | - | 8 - 9 | 1430 | 50 | |
| BC163513.1 | - | 8 - 9 | 1518 | 50 | ||
| BC163531.1 | - | 8 - 9 | 1518 | 50 | ||
More Information
Set Membership: |
|

angpt1
Genomic Map