Assay Details
| Gene Symbol: tnfsf10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131843.2 | NP_571918.2 | 3 - 4 | 435 | 130 | |
| GenBank mRNA | AF250041.1 | - | 2 - 3 | 130 | ||
| BC044336.1 | - | 3 - 4 | 435 | 130 | ||
| DQ812112.1 | - | 3 - 4 | 130 | |||
More Information
Set Membership: |
|

tnfsf10l
Genomic Map