Assay Details
| Gene Symbol: mafa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131844.2 | NP_571919.2 | 1 | 848 | 104 | |
| XM_017351895.2 | XP_017207384.1 | 1 | 1240 | 104 | ||
| XM_021467572.1 | XP_021323247.1 | 1 | 1240 | 104 | ||
| GenBank mRNA | AF109781.1 | - | 1 | 744 | 104 | |
| BC065941.1 | - | 1 | 837 | 104 | ||
| EH583521.1 | - | 1 | 880 | 103 | ||
More Information
Set Membership: |
|

mafa
Genomic Map