Assay Details
| Gene Symbol: gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131860.2 | NP_571935.2 | 7 - 8 | 1372 | 68 | |
| GenBank mRNA | BC163403.1 | - | 7 - 8 | 1372 | 68 | |
More Information
Set Membership: |
|

gpc4
Genomic Map