Assay Details
| Gene Symbol: sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153661.4 | NP_705947.3 | 2 - 3 | 196 | 69 | |
| GenBank mRNA | AY099515.2 | - | 2 - 3 | 187 | 69 | |
| BC056532.1 | - | 2 - 3 | 182 | 69 | ||
| BC067655.1 | - | 2 - 3 | 186 | 69 | ||
| EH501773.1 | - | 2 - 3 | 218 | 69 | ||
More Information
Set Membership: |
|

sf3b4
Genomic Map