Assay Details
| Gene Symbol: neo1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328497.1 | NP_001315426.1 | 27 - 28 | 4421 | 86 | |
| NM_173218.2 | NP_775325.2 | 27 - 28 | 4478 | 86 | ||
| XM_005166588.3 | XP_005166645.1 | 27 - 28 | 4487 | 86 | ||
| XM_009303464.3 | XP_009301739.1 | 27 - 28 | 4544 | 86 | ||
| XM_009303465.3 | XP_009301740.1 | 27 - 28 | 4542 | 86 | ||
| XM_009303466.3 | XP_009301741.1 | 26 - 27 | 4511 | 86 | ||
| XM_009303468.3 | XP_009301743.1 | 25 - 26 | 4293 | 86 | ||
| XM_017356809.2 | XP_017212298.1 | 26 - 27 | 4386 | 86 | ||
| XM_021477387.1 | XP_021333062.1 | 27 - 28 | 4545 | 86 | ||
| GenBank mRNA | AF394058.1 | - | 27 - 28 | 4289 | 86 | |
| AY029280.1 | - | 27 - 28 | 4426 | 86 | ||
| AY082380.1 | - | 27 - 28 | 4368 | 86 | ||
More Information
Set Membership: |
|

neo1a
Genomic Map