Assay Details
| Gene Symbol: neo1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328497.1 | NP_001315426.1 | 19 - 20 | 3251 | 66 | |
| NM_173218.2 | NP_775325.2 | 19 - 20 | 3308 | 66 | ||
| XM_005166588.3 | XP_005166645.1 | 19 - 20 | 3317 | 66 | ||
| XM_009303464.3 | XP_009301739.1 | 19 - 20 | 3374 | 66 | ||
| XM_009303465.3 | XP_009301740.1 | 19 - 20 | 3372 | 66 | ||
| XM_009303466.3 | XP_009301741.1 | 19 - 20 | 3374 | 66 | ||
| XM_009303468.3 | XP_009301743.1 | 19 - 20 | 3375 | 66 | ||
| XM_017356809.2 | XP_017212298.1 | 19 - 20 | 3375 | 66 | ||
| XM_021477387.1 | XP_021333062.1 | 19 - 20 | 3375 | 66 | ||
| GenBank mRNA | AF394058.1 | - | 19 - 20 | 3119 | 66 | |
| AY029280.1 | - | 19 - 20 | 3256 | 66 | ||
| AY082380.1 | - | 19 - 20 | 3198 | 66 | ||
| BC124155.1 | - | 19 - 20 | 3253 | 66 | ||
| CU458829.1 | - | 19 - 20 | 2946 | 66 | ||
More Information
Set Membership: |
|

neo1a
Genomic Map