Assay Details
| Gene Symbol: neo1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328497.1 | NP_001315426.1 | 20 - 21 | 3451 | 65 | |
| NM_173218.2 | NP_775325.2 | 20 - 21 | 3508 | 65 | ||
| XM_005166588.3 | XP_005166645.1 | 20 - 21 | 3517 | 65 | ||
| XM_009303464.3 | XP_009301739.1 | 20 - 21 | 3574 | 65 | ||
| XM_009303465.3 | XP_009301740.1 | 20 - 21 | 3572 | 65 | ||
| XM_009303468.3 | XP_009301743.1 | 20 - 21 | 3575 | 65 | ||
| XM_017356809.2 | XP_017212298.1 | 20 - 21 | 3575 | 65 | ||
| XM_021477387.1 | XP_021333062.1 | 20 - 21 | 3575 | 65 | ||
| GenBank mRNA | AY029280.1 | - | 20 - 21 | 3456 | 65 | |
| AY082380.1 | - | 20 - 21 | 3398 | 65 | ||
| BC124155.1 | - | 20 - 21 | 3453 | 65 | ||
| CU458829.1 | - | 20 - 21 | 3146 | 65 | ||
More Information
Set Membership: |
|

neo1a
Genomic Map