Assay Details
| Gene Symbol: pcdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182885.1 | NP_878305.1 | 3 - 4 | 3114 | 92 | |
| XM_009291096.3 | XP_009289371.1 | 3 - 4 | 3159 | 92 | ||
| GenBank mRNA | AB086625.1 | - | 3 - 4 | 3114 | 92 | |
| BC070019.1 | - | 3 - 4 | 2714 | 92 | ||
More Information
Set Membership: |
|

pcdh10b
Genomic Map