Assay Details
| Gene Symbol: pcdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182885.1 | NP_878305.1 | 4 - 5 | 3465 | 63 | |
| GenBank mRNA | AB086625.1 | - | 4 - 5 | 3465 | 63 | |
| BC070019.1 | - | 4 - 5 | 3065 | 63 | ||
More Information
Set Membership: |
|

pcdh10b
Genomic Map