Assay Details
| Gene Symbol: mmp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194416.1 | NP_919397.1 | 4 - 5 | 988 | 62 | |
| GenBank mRNA | AY324196.1 | - | 4 - 5 | 988 | 62 | |
| BC131864.1 | - | 4 - 5 | 980 | 62 | ||
More Information
Set Membership: |
|

mmp14a
Genomic Map