Assay Details
| Gene Symbol: evla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199519.3 | NP_955813.3 | 11 - 12 | 1462 | 120 | |
| XM_005158792.4 | XP_005158849.1 | 12 - 13 | 2088 | 120 | ||
| XM_005158793.4 | XP_005158850.1 | 12 - 13 | 2081 | 120 | ||
| XM_005158794.4 | XP_005158851.1 | 12 - 13 | 1876 | 120 | ||
| XM_005158795.4 | XP_005158852.1 | 12 - 13 | 2215 | 120 | ||
| XM_005158796.4 | XP_005158853.1 | 10 - 11 | 1792 | 120 | ||
| XM_005158797.4 | XP_005158854.1 | 11 - 12 | 1329 | 120 | ||
| XM_005158798.4 | XP_005158855.1 | 11 - 12 | 1327 | 120 | ||
| XM_005158799.4 | XP_005158856.1 | 11 - 12 | 1324 | 120 | ||
| XM_021467223.1 | XP_021322898.1 | 12 - 13 | 2086 | 120 | ||
| GenBank mRNA | BC056325.1 | - | 11 - 12 | 1433 | 120 | |
| BC065353.1 | - | 11 - 12 | 1426 | 120 | ||
More Information
Set Membership: |
|

evla
Genomic Map