Assay Details
| Gene Symbol: evla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199519.3 | NP_955813.3 | 3 - 4 | 742 | 68 | |
| XM_005158792.4 | XP_005158849.1 | 3 - 4 | 615 | 68 | ||
| XM_005158793.4 | XP_005158850.1 | 3 - 4 | 608 | 68 | ||
| XM_005158794.4 | XP_005158851.1 | 3 - 4 | 403 | 68 | ||
| XM_005158795.4 | XP_005158852.1 | 3 - 4 | 742 | 68 | ||
| XM_005158797.4 | XP_005158854.1 | 3 - 4 | 609 | 68 | ||
| XM_005158798.4 | XP_005158855.1 | 3 - 4 | 610 | 68 | ||
| XM_005158799.4 | XP_005158856.1 | 3 - 4 | 604 | 68 | ||
| XM_021467223.1 | XP_021322898.1 | 3 - 4 | 616 | 68 | ||
| GenBank mRNA | BC056325.1 | - | 3 - 4 | 713 | 68 | |
| BC065353.1 | - | 3 - 4 | 706 | 68 | ||
More Information
Set Membership: |
|

evla
Genomic Map