Assay Details
| Gene Symbol: syncripl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199679.1 | NP_955973.1 | 4 - 5 | 477 | 64 | |
| XM_005158913.4 | XP_005158970.1 | 3 - 4 | 383 | 64 | ||
| XM_009293300.3 | XP_009291575.1 | 4 - 5 | 550 | 64 | ||
| GenBank mRNA | AY398389.1 | - | 4 - 5 | 568 | 64 | |
| BC046902.1 | - | 4 - 5 | 477 | 64 | ||
| BC066570.1 | - | 4 - 5 | 509 | 64 | ||
More Information
Set Membership: |
|

syncripl
Genomic Map