Assay Details
| Gene Symbol: syncripl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199679.1 | NP_955973.1 | 5 - 6 | 582 | 95 | |
| XM_005158913.4 | XP_005158970.1 | 4 - 5 | 488 | 95 | ||
| XM_009293300.3 | XP_009291575.1 | 5 - 6 | 655 | 95 | ||
| GenBank mRNA | AY398389.1 | - | 5 - 6 | 673 | 95 | |
| BC046902.1 | - | 5 - 6 | 582 | 95 | ||
| BC066570.1 | - | 5 - 6 | 614 | 95 | ||
More Information
Set Membership: |
|

syncripl
Genomic Map