Assay Details
| Gene Symbol: syncripl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199679.1 | NP_955973.1 | 7 - 8 | 893 | 66 | |
| XM_005158913.4 | XP_005158970.1 | 6 - 7 | 799 | 66 | ||
| XM_009293300.3 | XP_009291575.1 | 7 - 8 | 966 | 66 | ||
| GenBank mRNA | AY398389.1 | - | 7 - 8 | 984 | 66 | |
| BC046902.1 | - | 7 - 8 | 893 | 66 | ||
| BC066570.1 | - | 7 - 8 | 925 | 66 | ||
More Information
Set Membership: |
|

syncripl
Genomic Map