Assay Details
| Gene Symbol: snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199694.1 | NP_955988.1 | 4 - 5 | 569 | 66 | |
| XM_021470452.1 | XP_021326127.1 | 4 - 5 | 631 | 66 | ||
| GenBank mRNA | BC062858.1 | - | 4 - 5 | 569 | 66 | |
More Information
Set Membership: |
|

snupn
Genomic Map