Assay Details
| Gene Symbol: larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199930.2 | NP_956224.1 | 5 - 6 | 693 | 75 | |
| XM_005166534.4 | XP_005166591.1 | 5 - 6 | 659 | 75 | ||
| GenBank mRNA | BC049455.1 | - | 5 - 6 | 695 | 75 | |
More Information
Set Membership: |
|

larp7
Genomic Map