Assay Details
| Gene Symbol: wasb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199938.1 | NP_956232.1 | 10 - 11 | 1395 | 83 | |
| GenBank mRNA | BC053291.1 | - | 10 - 11 | 1395 | 83 | |
More Information
Set Membership: |
|

wasb
Genomic Map