Assay Details
| Gene Symbol: wasb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199938.1 | NP_956232.1 | 6 - 7 | 615 | 73 | |
| GenBank mRNA | BC053291.1 | - | 6 - 7 | 615 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wasb| Gene Symbol: wasb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199938.1 | NP_956232.1 | 6 - 7 | 615 | 73 | |
| GenBank mRNA | BC053291.1 | - | 6 - 7 | 615 | 73 | |
Set Membership: |
|