Assay Details
| Gene Symbol: wasb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199938.1 | NP_956232.1 | 9 - 10 | 1000 | 76 | |
| GenBank mRNA | BC053291.1 | - | 9 - 10 | 1000 | 76 | |
More Information
Set Membership: |
|

wasb
Genomic Map