Assay Details
| Gene Symbol: lmbrd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199944.1 | NP_956238.1 | 10 - 11 | 1406 | 74 | |
| GenBank mRNA | BC063324.1 | - | 10 - 11 | 1406 | 74 | |
More Information
Set Membership: |
|

lmbrd2b
Genomic Map