Assay Details
| Gene Symbol: chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200045.1 | NP_956339.1 | 11 - 12 | 1298 | 65 | |
| XM_005165030.4 | XP_005165087.1 | 11 - 12 | 65 | |||
| GenBank mRNA | AF265346.1 | - | 11 - 12 | 1266 | 65 | |
| BC044519.1 | - | 11 - 12 | 1298 | 65 | ||
More Information
Set Membership: |
|

chek2
Genomic Map