Assay Details
| Gene Symbol: lipg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200128.1 | NP_956422.1 | 9 - 10 | 1597 | 61 | |
| GenBank mRNA | BC044146.1 | - | 9 - 10 | 1597 | 61 | |
More Information
Set Membership: |
|

lipg
Genomic Map