Assay Details
| Gene Symbol: reep3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200161.1 | NP_956455.1 | 3 - 4 | 449 | 111 | |
| GenBank mRNA | BC045373.1 | - | 3 - 4 | 449 | 111 | |
More Information
Set Membership: |
|

reep3b
Genomic Map