Assay Details
| Gene Symbol: rasl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200395.1 | NP_956689.1 | 4 - 5 | 413 | 80 | |
| XM_017356827.2 | XP_017212316.1 | 4 - 5 | 607 | 80 | ||
| GenBank mRNA | BC054128.1 | - | 4 - 5 | 413 | 80 | |
More Information
Set Membership: |
|

rasl12
Genomic Map