Assay Details
| Gene Symbol: hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200723.1 | NP_957017.1 | 2 - 3 | 317 | 96 | |
| XM_021468015.1 | XP_021323690.1 | 1 - 2 | 181 | 96 | ||
| GenBank mRNA | BC059489.1 | - | 2 - 3 | 317 | 96 | |
More Information
Set Membership: |
|

hpca
Genomic Map