Assay Details
| Gene Symbol: tbc1d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200777.1 | NP_957071.1 | 6 - 7 | 868 | 60 | |
| XM_021468380.1 | XP_021324055.1 | 6 - 7 | 897 | 60 | ||
| GenBank mRNA | BC059618.1 | - | 6 - 7 | 868 | 60 | |
More Information
Set Membership: |
|

tbc1d7
Genomic Map