Assay Details
| Gene Symbol: camk1gb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200829.1 | NP_957123.1 | 6 - 7 | 628 | 100 | |
| XM_005174131.4 | XP_005174188.2 | 6 - 7 | 693 | 100 | ||
| XM_005174132.4 | XP_005174189.1 | 6 - 7 | 771 | 100 | ||
| XM_021469766.1 | XP_021325441.1 | 6 - 7 | 738 | 100 | ||
| GenBank mRNA | BC060911.1 | - | 6 - 7 | 628 | 100 | |
More Information
Set Membership: |
|

camk1gb
Genomic Map