Assay Details
| Gene Symbol: hsd17b12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200881.1 | NP_957175.1 | 10 - 11 | 972 | 82 | |
| GenBank mRNA | AY551082.1 | - | 10 - 11 | 965 | 82 | |
| BC048053.1 | - | 10 - 11 | 914 | 82 | ||
| BC063943.1 | - | 10 - 11 | 972 | 82 | ||
| BC164345.1 | - | 10 - 11 | 889 | 82 | ||
More Information
Set Membership: |
|

hsd17b12a
Genomic Map